Anti-CXCR1 Mouse Monoclonal Antibody (Fluorescein) [clone: 42705]

Supplier: R&D Systems
FAB330F-025 FAB330F-100
102882-456EA 172.09 USD
102882-456 102882-458
Anti-CXCR1 Mouse Monoclonal Antibody (Fluorescein) [clone: 42705]
Antibodies
Type: Primary
Antigen: CXCR1
Clonality: Monoclonal
Clone: 42705
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR