Anti-KIR3DL2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 539304]

Supplier: R&D Systems
FAB2878A-025 FAB2878A-100
102882-306EA 201.02 USD
102882-306 102882-308
Anti-KIR3DL2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 539304]
Antibodies
Type: Primary
Antigen: KIR3DL2
Clonality: Monoclonal
Clone: 539304
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR