Anti-CCR10 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 248918]

Supplier: R&D Systems
FAB2815C-025 FAB2815C-100
102882-010EA 215.48 USD
102882-010 102882-012
Anti-CCR10 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 248918]
Antibodies
Type: Primary
Antigen: CCR10
Clonality: Monoclonal
Clone: 248918
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR