Anti-KIR2DL4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 181703]

Supplier: R&D Systems
FAB2238P-025 FAB2238P-100
102881-924EA 186.55 USD
102881-924 102881-926
Anti-KIR2DL4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 181703]
Antibodies
Type: Primary
Antigen: KIR2DL4
Clonality: Monoclonal
Clone: 181703
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR