Anti-CXCR6 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 221002]

Supplier: R&D Systems
FAB2145P-025 FAB2145P-100
102881-814EA 186.55 USD
102881-814 102881-816
Anti-CXCR6 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 221002]
Antibodies
Type: Primary
Antigen: CXCR6
Clonality: Monoclonal
Clone: 221002
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR