Anti-CXCR6 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 221002]

Supplier: R&D Systems
FAB2145A-025 FAB2145A-100
102881-798EA 215.48 USD
102881-798 102881-800
Anti-CXCR6 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 221002]
Antibodies
Type: Primary
Antigen: CXCR6
Clonality: Monoclonal
Clone: 221002
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR