Anti-KIR2DL3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 180701]

Supplier: R&D Systems
FAB2014P-025 FAB2014P-100
102881-682EA 215.48 USD
102881-682 102881-684
Anti-KIR2DL3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 180701]
Antibodies
Type: Primary
Antigen: KIR2DL3
Clonality: Monoclonal
Clone: 180701
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR