Anti-IL12RB1 Goat Polyclonal Antibody (PE (Phycoerythrin))

Supplier: R&D Systems
FAB1998P-025 FAB1998P-100
102881-668EA 273.32 USD
102881-668 102881-670
Anti-IL12RB1 Goat Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: IL12RB1
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR