Anti-IL12RB2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 305719]

Supplier: R&D Systems
FAB1959P-025 FAB1959P-100
102881-600EA 302.25 USD
102881-600 102881-602
Anti-IL12RB2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 305719]
Antibodies
Type: Primary
Antigen: IL12RB2
Clonality: Monoclonal
Clone: 305719
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR