Anti-CXCR5 Mouse Monoclonal Antibody (Fluorescein) [clone: 51505]

Supplier: R&D Systems
FAB190F-025 FAB190F-100
102881-382EA 215.48 USD
102881-382 102881-384
Anti-CXCR5 Mouse Monoclonal Antibody (Fluorescein) [clone: 51505]
Antibodies
Type: Primary
Antigen: CXCR5
Clonality: Monoclonal
Clone: 51505
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR