Anti-CCR9 Mouse Monoclonal Antibody (Fluorescein) [clone: 112509]

Supplier: R&D Systems
FAB179F-025 FAB179F-100
102881-444EA 215.48 USD
102881-444 102881-446
Anti-CCR9 Mouse Monoclonal Antibody (Fluorescein) [clone: 112509]
Antibodies
Type: Primary
Antigen: CCR9
Clonality: Monoclonal
Clone: 112509
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR