Anti-CCR2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 48607]

Supplier: R&D Systems
FAB151P-025 FAB151P-100
102880-526EA 258.86 USD
102880-526 102880-528
Anti-CCR2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 48607]
Antibodies
Type: Primary
Antigen: CCR2
Clonality: Monoclonal
Clone: 48607
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR