Anti-MICA Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 159227]

Supplier: R&D Systems
FAB1300G-025 FAB1300G-100
102881-152EA 244.4 USD
102881-152 102881-154
Anti-MICA Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 159227]
Antibodies
Type: Primary
Antigen: MICA
Clonality: Monoclonal
Clone: 159227
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR