Anti-KLRC1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 131411]

Supplier: R&D Systems
FAB1059A-025 FAB1059A-100
102880-170EA 273.32 USD
102880-170 102880-172
Anti-KLRC1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 131411]
Antibodies
Type: Primary
Antigen: KLRC1
Clonality: Monoclonal
Clone: 131411
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR