Glycogen, powder

Supplier: SPECTRUM CHEMICAL MFG CORP

GL160-25GM GL160-100GM GL160-1GM GL160-500GM
700013-198EA 751.65 USD
700013-198 700013-202 700013-194 700013-196 700013-200
Glycogen, powder
Glycogen

Glycogen, Powder is a multibranched polysaccharide of glucose which represent the primary storage of glucose in the body, as well as serving as an energy storage in fungi and animals.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₄H₄₂O₂₁
MW: 666.579 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00081547
CAS Number: 9005-79-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR