Polyethylene glycol 3,350, Biotechnology Grade for biotechnology

Supplier: VWR

Synonyms: PEG 3,350, PEG 3350

0955-500G 0955-1KG
10791-816CS 879.63 USD
10791-816 10791-818
Polyethylene glycol 3,350, Biotechnology Grade for biotechnology
Polyethylene glycol 3,350
Formula: H(OCH₂CH₂)nOH
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now

Specification Test Results

pH (5%, Water, 25 °C) 4.5 - 7.5
Viscosity Range 76 - 110 cS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR