Ear Punch, Stoelting
Supplier: Stoelting
56718 56719
100231-754EA 65.38 USD
100231-754 100231-756
Ear Punch, Stoelting
Biopsy Punches
Ear punches used for motching or punch in mouse or rat ears at weaning for easy identification.
  • Made of Stainless steel
  • Ergonomic handles for easy control
  • Non-clinical use only
Ear punches used for motching or punch in mouse or rat ears at weaning for easy identification. Made of Stainless steel, with ergonomic handles for easy control. For non-clinical use only.