Ribbon (Slepyan) Stitch Scissors, OR Grade, Sklar
Supplier: Sklar
64-1445 64-1345
10194-142EA 192.74 USD
10194-142 10197-858
Ribbon (Slepyan) Stitch Scissors, OR Grade, Sklar
Scissors
Ribbon Stitch Scissors are used for suture removal
These scissors have short blades with sharp, fine tips, that are used to snip the suture rather than lifting the suture slightly first, as with Spencer or Littauer stitch scissors.
These Stitch Scissors should not be used for cutting tissue, but may be useful for delicate or micro-dissection. They also feature Ribbon Style Finger Ring handles for increased comfort and improved hold.