NIST Polystyrene Test Film, International Crystal Labs
Supplier: International Crystal Labs
0009-2500 0009-7676
100213-626EA 58.46 USD
100213-626 100215-952
NIST Polystyrene Test Film, International Crystal Labs
Spectrophotometers and Photometers UV/Visible Spectrophotometers
Films are available in free standing pairs mounted on 2×4" cards that fit in the standard universal slide mount in all spectrophotometers or the films can be cast on KBr windows.
Films cast on KBr windows will not exhibit any interference fringes.