Crile-Murray Needle Holder, OR Grade, Sklar
Supplier: Sklar
20-1260 21-1260
82030-088EA 142.59 USD
82030-088 89023-042
Crile-Murray Needle Holder, OR Grade, Sklar
Forceps
Crile-Murray Needle Holder is used to hold and guide small to medium size needles and suture materials. The ratcheted finger ring handle aids in securing needles for both continuous and interrupted closures. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.
The Crile-Murray Needle Holder is comparable to the Crile-Wood Needle Holder.
Censitrac Ready products are industry standard and constructed of stainless steel.
Información para pedidos: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Precaución: Customers must have a Censitrac Site ID to purchase Censitrac instruments.