TC Mathieu Needle Holder, OR Grade, Sklar®
Supplier: Sklar
21-8070 21-8072 21-8071 21-8074 21-8076 21-8077 21-8078 21-8073
89023-398EA 379.26 USD
89023-398 10195-192 75817-766 76192-666 76192-668 76192-670 76192-672 76192-736
TC Mathieu Needle Holder, OR Grade, Sklar®
Forceps
The Sklar® Mathieu TC Needle Holder is a single spring instrument with a plier handle grip that curves inward at the bottom and features a ratchet locking mechanism at the base of the handles that can be easily squeezed to be locked and unlocked.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
This needle holder is used when more refined suturing is required. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles.