TC Olsen-Hegar Needle Holder, OR Grade, Sklar
Supplier: Sklar
21-8025 21-8026 21-8027 21-8028 21-8029
89023-346EA 376.38 USD
89023-346 89023-348 89023-350 89023-352 89023-354
TC Olsen-Hegar Needle Holder, OR Grade, Sklar
Forceps
Olsen-Hegar TC Needle Holder is a ratcheted, finger ring instrument with built in scissors
  • Censitrac Ready products are industry standard and constructed of stainless steel
  • Latex-free
Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.
It is used with intermediate weight sutures and in plastic surgery procedures.
Información de suministro: Ordering Information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Precaución: Customers must have a Censitrac Site ID to purchase Censitrac instruments.