TC Halsey Needle Holder, OR Grade, Sklar®
Supplier: Sklar
21-8000 21-8001 21-8098 21-8099
89023-306EA 274.74 USD
89023-306 89023-308 76192-690 76192-692
TC Halsey Needle Holder, OR Grade, Sklar®
Forceps
Sklar® TC Halsey Needle Holders are ratcheted, used for holding small needles and fine suture material which is commonly used in plastic surgery or subcutaneous procedures around the face, hands, and delicate tissue, such as dermatology.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
TC Halsey Needle Holders are available with smooth or delicately serrated jaws. Smooth jaws will cause the least damage to the needle, however serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized. Sklar® permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.