Pierce™ Kinase Enrichment Kit, Thermo Scientific
Supplier: LIFE TECHNOLOGIES MS
The Pierce™ Kinase Enrichment Kits utilise an ActivX™ ATP or ADP probe to covalently label the active site of ATPases, including chaperones and metabolic enzymes, to enable their selective enrichment using a desthiobiotin tag.
- Labels Only the Conserved Active-Site Lysines of Nucleotide-Binding Proteins
- Flexible use for in vitro Labeling of ATPase Enzymes Derived from Cells or Tissues
- Compatible with use with Western Blot or Mass Spectrometry (MS) Workflows
ActivX™ probes feature an amine-reactive nucleotide analog and a desthiobiotin (biotin analogue) tag that facilitates selective labeling of lysines in the kinase active site and then subsequent enrichment and recovery of labelled protein.
Precaución: For Research Use Only. Not for use in diagnostic procedures.