Pierce™ High Sensitivity NeutrAvidin™ HRP
Supplier: LIFE TECHNOLOGIES MS
31030 31032
PI31030EA 306.46 USD
PI31030 PI31032
Pierce™ High Sensitivity NeutrAvidin™ HRP
Proteins and Peptides
These High Sensitivity NeutrAvidin HRP Conjugate is specially prepared peroxidase-conjugated form of avidin biotin-binding protein that provides signal amplification like poly-HRP and exceptional storage stability.
  • High specificity, low background binding
  • High sensitivity which detects low levels of target without background; obtain high signal-to-noise ratios
  • Cost effective, uses less conjugate in Western blotting and ELISA applications while still obtaining excellent results
  • Compatible with typical chemiluminescent, fluorescent and colorimetric peroxidase substrates
  • Ready-to-use stabilised liquid format means there is no waiting for thawing and no need to aliquot
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.
Precaución: For Research Use Only. Not for use in diagnostic procedures.