Pierce™ Glutathione Chromatography Cartridges, Thermo Scientific
Supplier: Invitrogen
16109 16110
PI16109PK 453.71 USD
PI16109 PI16110
Pierce™ Glutathione Chromatography Cartridges, Thermo Scientific
Chromatography Columns
Thermo Scientific Pierce Glutathione Chromatography Cartridges are convenient, ready-to-use pre-packed devices for the purification of GST-fusion proteins from cellular lysates.
  • Binds at least 40 mg of purified recombinant GST protein per ml of resin
  • Consistently purifies at least 10 mg of GST-tagged protein per mL of resin with greater than 90% purity
  • Resin can be reused at least five times without reduction in binding capacity and purification performance
  • Works well to purify GST-fusion proteins from bacterial lysates or use with pre-purified GST-tagged proteins to pull down protein interactions
Pierce™ Glutathione chromatography cartridges are compatible with the major automated liquid-chromatography systems or manual syringe processing. The cartridges attach directly to ÄKTA™ or FPLC Systems without additional connectors. The included accessory pack readily adapts cartridges for use with Luer-Lok® syringe fittings or ¹/₁₆" tubing. These cartridges enable fast, easy and reproducible chromatographic separations and can be regenerated for multiple uses.
Información de suministro: Kit includes an accessory pack (1 female Luer-Lok® adapter, 1 connector fitting, 1 column plug and bottom caps).
Precaución: For Research Use Only. Not for use in diagnostic procedures.