AlcoSCRUB Foaming Hand Sanitizer, Thermo Scientific
Supplier: Erie Scientific
43032-08-001 43002-24-001
89094-988CS 313.35 USD
89094-988 89094-986
AlcoSCRUB Foaming Hand Sanitizer, Thermo Scientific
Skincare Skin Disinfectants Liquid and Foam Skin Disinfectants
AlcoSCRUB Foaming Hand Sanitizer is an effective antiseptic hand cleanser to kill harmful bacteria.
AlcoSCRUB Foam contains 62% ethyl alcohol and is 99.9% effective against a broad spectrum of microorganisms. No sticky residue; formula absorbs completely.
Información para pedidos: A wall bracket is available as a separate accessory. The bracket mounts to the wall using two-sided tape, and the bottles can be easily snapped into the dispenser.