Equipment and Moving Caution Signs, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
C130AB C130P C130PB C130R C130RB C136P C136PB C136R C136RB C146P C146PB C146R C146RB C352R C352RB C383AP C398P C398PB C398R C398RB C450AB C450PB C450RB C478AB C478PB C478RB C489AB C489PB C489RB C490AB C490PB C490RB C500AB C500PB C500RB C551AB C551PB C551RB C559AB C559PB C559RB C563AB C563AC C563AD C563PB C563RB C563RC C563RD C571AB C571PB C571RB C572AB C572PB C572RB C573AB C573PB C573RB C576AB C576PB C576RB C577AB C577PB C577RB C636AB C636PB C636RB ESC130AB ESC130PB ESC130RB SPSA122P SPSA122R
94004-704EA 51.81 USD
94004-704 94004-706 94004-708 94004-710 94004-712 94004-726 94004-728 94004-730 94004-732 94004-758 94004-760 94004-762 94004-764 94005-130 94005-132 94005-310 94005-414 94005-416 94005-418 94005-420 94006-042 94006-044 94006-046 94006-210 94006-212 94006-214 94006-276 94006-278 94006-280 94006-282 94006-284 94006-286 94006-360 94006-362 94006-364 94006-660 94006-662 94006-664 94006-718 94006-720 94006-722 94006-754 94006-756 94006-758 94006-760 94006-762 94006-764 94006-766 94006-826 94006-828 94006-830 94006-832 94006-834 94006-836 94006-838 94006-840 94006-842 94006-856 94006-858 94006-860 94006-862 94006-864 94006-866 94007-262 94007-264 94007-266 94013-088 94013-090 94013-092 94021-876 94021-878
Equipment and Moving Caution Signs, National Marker
Signs
OSHA signs are the perfect way to alert your employees and guests when a hazard is present in your facility.
  • Ideal for indoor and outdoor labeling
  • Available in Alum, PS vinyl and Rigid plastic (thick polystyrene) materials
  • .040 alum is weatherproof, non-corrosive and has baked-on enamel on both sides
  • .004 PS vinyl material has pressure sensitive adhesive backing
  • .050 rigid plastic material has radius corners and pre-drilled corner mounting holes
Keep your facility safe, buy an OSHA regulated sign today and prevent an accident tomorrow. Each sign header reads, "Cautiion".
Certificaciones: Comply with OSHA 29 CFR 1910.145