MS-275 ≥95% (by 1H-NMR)
Supplier: Enzo Life Sciences
Synonyms:
Pyridin-3-ylmethyl 4-((2-aminophenyl)carbamoyl)benzylcarbamate, Entinostat
270-378-M025
270-378-M001
89147-740EA
294
USD
89147-740
89147-742
76001-974
MS-275 ≥95% (by 1H-NMR)
MS-275
HDAC1 inhibitor
Preferentially inhibits HDAC1 (IC50=300nM) over HDAC3 (IC50=8µM). Has no inhibitory activity towards HDAC8 (IC50>100µM).
Formula:
C₂₁H₂₀N₄O₃ MW: 376.4 g/mol |
MDL Number:
MFCD08272435 CAS Number: 209783-80-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...