Universal Immunoassay Buffer, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

25880-01 25880-10
102092-354EA 90.31 USD
102092-354 102092-356
Universal Immunoassay Buffer, Electron Microscopy Sciences
Buffers Assay Buffers

Immunoassay buffer is used as a 1X in all immunoassays (immunofluorescence, IHC, ELISA, WB etc.) as a washing buffer and it can be used as a dilution buffer for antibodies; however it does not contain any carrier proteins.


  • Buffer also does not contain any phosphate, sodium azide or mercury as preservatives
  • Can be used for dilution of all antibodies, including peroxidase, antibodies to phosphoproteins
  • Not suitable for dilution of antibodies to S100 proteins


Application: Dilute this 10X buffer 10 times with DI water. (90 ml water + 10 ml this buffer – mixed well). Wash 3-5 times with this buffer in between the steps of your immunoassay protocol.


Información de suministro: Supplied as 10X buffer.


Precaución: Store at room temperature.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR