Uranyl magnesium acetate
Supplier: Electron Microscopy Sciences
|
Danger
|
22501
22500
101410-838EA
143.71
USD
101410-838
101410-836
Uranyl magnesium acetate
Uranyl magnesium acetate
Uranyl magnesium acetate can be used as a substitute for uranyl acetate in either double-staining techniques and also as a solo stain.
Formula:
UO₂Mg(CH₃COO)₄ Storage Temperature: Ambient |
MDL Number:
MFCD00036427 CAS Number: 20596-93-4 UN: 2910 ADR: 7, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...