Uranyl formate 99.9-100.0%

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Uranium, bis(formato-kappao)dioxo-, (T-4)-, bis(formato-O)dioxouranium

22450 22451
101410-832EA 219.15 USD
101410-832 101410-834
Uranyl formate 99.9-100.0%
Uranyl formate

Uranyl formate offers amazing advancement in negative staining for microscopy.


  • For ultra fine grain structure

Formula: C₂H₂O₆U
MW: 360.06 g/mol
Melting Pt: 110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041939
CAS Number: 16984-59-1
UN: 2910
ADR: 7

Order Now

Specification Test Results

Purity 99.9-100%
Specific gravity 3.695


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR