Maximow’s Method for Bone Marrow, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
26252-01 26252-03 26252-04 26252-02
101411-198EA 80.6 USD
101411-198 101411-202 101411-204 101411-200
Maximow’s Method for Bone Marrow, Electron Microscopy Sciences
Stains Histology Stains
Maximow’s method for bone marrow is used for morphology details of marrow cells.
  • Formalin fixation
  • Paraffin at 6 microns