Russell's Zenker Fixative, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
64123-05 64123-06
101412-888EA 51.66 USD
101412-888 101412-890
Russell's Zenker Fixative, Electron Microscopy Science
Stains Microscopy and staining reagents
Russell's Zenker Fixative replacement for Zenker's Stock solution.
  • Use in the same manner as Zenker's stock solution
  • Comparable results to Zenker's Stock solution
  • Contains Zinc Chloride
Due to the hazardous nature of mercury, Zenker’s fluid is no longer available. In place we are offering Russell’s Zenker Fixative, which contains Zinc Chloride. The staining results produced by this fixative are comparable to those of Zenker’s Stock solution. Russell’s Modified Zenker’s is used in the same manner as Zenker’s stock solution.