Lillie’s Method for Iron, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
26604-01 26604-02 26604-03 26604-04 26604-1A
101411-426EA 70.73 USD
101411-426 101411-428 101411-430 101411-432 101411-434
Lillie’s Method for Iron, Electron Microscopy Sciences
Stains Histology Stains
Stain results - Ferric Iron: Dark Prussian Blue, Ferrous Iron: Dark Turnbull’s Blue, Background: Light Red.