Gridley’s Method for Endamoeba Histolytica, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
26109-01
26109-04
26109-05
26109-1B
26109-03
26109-1A
26109-02
101411-046EA
59.82
USD
101411-046
101411-052
101411-054
101411-058
101411-050
101411-056
101411-048
Gridley’s Method for Endamoeba Histolytica, Electron Microscopy Sciences
Stains
Histology Stains
Gridley’s Method for Endamoeba Histolytica
For bacteria stain: Amoebae, their nuclei and ingested erythrocytes. Stain results - Erythrocytes (ingested): Rose, Connective tissue: Green, Amoebae: Blue-green, Nuclei (amoebic): Darker blue-green.