Crystal Clear™ Water Liquid Plastics, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
24200 24204 24206
102091-016EA 594.32 USD
102091-016 102091-018 102091-020
Crystal Clear™ Water Liquid Plastics, Electron Microscopy Science
Stains Microscopy and staining reagents
The Crystal Clear™ series of polyurethane resins are designed for applications that require absolute clarity
  • Cured products are UV resistant and are not brittle
  • Low viscosity resins that cure at room temperature with negligible shrinkage to a hard plastic
The Crystal Clear™ Series of polyurethane resins were designed for applications that require absolute clarity. These are low viscosity resins that cure at room temperature with negligible shrinkage to a hard plastic. Cured products are UV resistant and are not brittle. Applications include potting, encapsulating, making prototype models, and lenses, object duplicating etc.
CAUTION: This product is NOT for home use, for research and industrial use only.
Each kit consists of 1 gallon resin A and adequate curing agent B
Información de suministro: Each kit consists of 1 gallon resin A and adequate curing agent B.
Precaución: This product is not for home use, for research and industrial use only.