Sodium hydroxide 2 N in aqueous solution

Supplier: Electron Microscopy Sciences

21170-01 21170
100504-376EA 30.89 USD
100504-376 100504-964
Sodium hydroxide 2 N in aqueous solution
Sodium hydroxide

2N Standardized and Carbonate-free. This solution is prepared from ACS Reagent grade sodium hydroxide, and is standardized colormetrically against potassium biphthalate primary reference material.
Normal Concentration 1.98-2.02

Formula: NaOH
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR