Sodium hydroxide ≥97.0%, pellets ACS

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Caustic soda

21160 21162
100504-282EA 16.73 USD
100504-282 100504-284
Sodium hydroxide ≥97.0%, pellets ACS
Sodium hydroxide

NaOH F.W.40.00 Assay 97.0%

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C (2534 °F)
Melting Pt: 318 °C (604 °F)
Density: 2.13 g/cm³ (17.775 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II

Order Now

Specification Test Results

Ammonium hydroxide 0.02% ppt
Assay >97.0%
Chloride 0.005%
Heavy metals 0.002%
Iron 0.001%
Mercury 0.1 ppm
Nickel 0.001%
Nitrogen compounds 0.001%
Phosphate 0.001%
Potassium 0.02%
Sodium carbonate 1%
Sulfate 0.003%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR