Schiff reagent
Supplier: Electron Microscopy Sciences
|
Danger
|
26052-05
26052-06
100504-432EA
100.09
USD
100504-432
100504-434
Schiff reagent
Schiff reagent
For use in the Periodic Acid Leucofuchsin (PAS) method as a general tissue stain.
- High quality staining solution
- Ready-to-use
Storage Temperature:
Refrigerator |
MDL Number:
MFCD00081985 UN: 1789 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...