di-Potassium oxalate monohydrate 98.5-101.0% ACS
Supplier: Electron Microscopy Sciences
|
Warning
|
Synonyms:
Oxalic acid potassium salt monohydrate, Potassium oxalate hydrate, Potassium oxalate monohydrate, Potassium ethanedioate monohydrate
20160
20162
100504-236EA
41.72
USD
100504-236
100504-238
di-Potassium oxalate monohydrate 98.5-101.0% ACS
di-Potassium oxalate monohydrate
(CO2K)2·H2O F.W. 184.23 CAS 6487-48-5
Assay: 98.5-101.0%
Neutrality: To Pass Test
Formula:
(COOK)₂·H₂O MW: 184.23 g/mol Density: 2.127 g/cm³ (17.75 lbs/gal.) Storage Temperature: Ambient |
MDL Number:
MFCD00150033 CAS Number: 6487-48-5 UN: 3282 ADR: 6.1,III |
Specification Test Results
Ammonium | 0.002% |
Assay | 98.5-101.0% |
Chloride | 0.002% |
Heavy metals | 0.0025% |
Insoluble matters | 0.01% |
Iron | 0.001% |
Neutrality | To pass test |
Sodium | 0.02% |
Sulfate | 0.01% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...