PIPES 1.5 sodium salt, crystals

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Piperazine-N,N-bis (2-ethanesulphonic acid) 1.5 Sodium salt

19240 19230
100504-180EA 290.92 USD
100504-180 100504-178
PIPES 1.5 sodium salt, crystals
PIPES 1.5 sodium salt

[Piperazine-N,N-bis (2-ethanesulfonic acid,) 1.5 Sodium] F.W. 335.3 pK = pH = 6.8 25°C CAS 10010-67-0

Storage Temperature: Ambient
CAS Number: 10010-67-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3aab57950c8e5500ba2e68e406611fc156018f0a&vid=49c8e144ed8fa0fef1fed3172f2727409edfa594&prid=web&clid=SAPDAM

Order Now

Specification Test Results

pH (25 °C) 6.8

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR