Nonenylsuccinic anhydride (mixture of branched chain isomers)

Supplier: Electron Microscopy Sciences
Warning

Synonyms: NSA, Isononenylsuccinic anhydride

19050 19051
100504-956EA 61.91 USD
100504-956 100504-958
Nonenylsuccinic anhydride (mixture of branched chain isomers)
Nonenylsuccinic anhydride (mixture of branched chain isomers)

An epoxy hardener specially distilled EM Grade. The special distillation process offers much clearer blocks than the standard NSA.

Formula: C₁₃H₂₀O₃
Boiling Pt: 325 °C
Density: 1.08 gm/cm³
Flash Pt: 13 °C (55 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014552
CAS Number: 28928-97-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9e062994d165a98713003c252ad401f7d5602f6&vid=a9e062994d165a98713003c252ad401f7d5602f6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Viscosity 102.8 at 25 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR