Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers)
Supplier: Electron Microscopy Sciences
|
Danger
|
19000
19001
100504-116EA
47.1
USD
100504-116
100504-118
Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers)
Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers)
An epoxy hardener.
Formula:
C₁₀H₁₀O₃ MW: 178.19 g/mol Boiling Pt: 140 °C (10 mm Hg) Density: 1.23 g/cm³ (10.264 lbs/gal.) Storage Temperature: Ambient |
MDL Number:
MFCD00167036 CAS Number: 25134-21-8 UN: 1760 ADR: 8,III |
Specification Test Results
Specific gravity at 20/20 °C | 1.245 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...