Metanil yellow stock solution 0,25%

Supplier: Electron Microscopy Sciences

Synonyms: 3-(4-Anilinophenylazo)benzenesulphonic acid sodium salt, Acid Yellow 36

26081-06 26081-05
100504-468EA 81.3 USD
100504-468 100504-466
Metanil yellow stock solution 0,25%
Metanil yellow
Formula: C₁₈H₁₄N₃NaO₃S
MDL Number: MFCD00007486
CAS Number: 587-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR