Hexamethylenetetramine 3% in aqueous solution

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Hexamethylenetetramine, Hexamine, Urotropine, Formin

26083-05 26083-06
100504-470EA 52.03 USD
100504-470 100504-884
Hexamethylenetetramine 3% in aqueous solution
Hexamethylenetetramine

A stock solution for use in bacterial and fungal stains (Grocott's Method/GMS).


  • High quality staining solution
  • Ready-to-use

Formula: C₆H₁₂N₄
MW: 140.19 g/mol
Density: 1.00993 g/cm³ (8.428 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00006895
CAS Number: 100-97-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26d39e1d753c53d55a7c78f7618c533166eb2972&vid=8b13241a362340ecbd4f79bf3f1871907674ebc2&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR