Methanol ≥99.8% ACS

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

18511 18510
100504-100EA 117.65 USD
100504-100 100504-098
Methanol ≥99.8% ACS
Methanol

(Methyl Alcohol) CH3OH F.W. 32.04 Assay: 99.8% Specific Gravity : 0.79 A.C.S.

Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6
Melting Pt: -98
Density: 0.79 g/cm³ (6.593 lbs/gal.)
Flash Pt: 11 °C (52 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
UN: 1230
ADR: 3 II
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17f87f215a11a98f9bcb01f5252a3fd11c9bfbbf&vid=ed098825e1273d8f5afcc4d963acb6aaedb21174&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.8%
Specific gravity 0.79
Residue after evaporation 0.001%
Boiling Range Max. 2 °C
Acetone and Aldehydes (0.001%) To Pass Test
Titratable Acid 0.0003 meq/g
Titratable Base 0.0002 meq/g
Water Max. 0.2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR