Mammalian Ringer solution
Supplier: Electron Microscopy Sciences
Warning
11763-05 11763-10
100496-410EA 56.54 USD
100496-410 100502-384
Mammalian Ringer solution
Mammalian ringer solution
Ingredients: NaCl; KCl; CaCl₂·6H₂O; NaHCO₃; MgCl₂·6H₂O; glucose and distilled water.