Lead citrate trihydrate ≥99%
Supplier: Electron Microscopy Sciences
|
|
The most widely used metal stain for ultra thin sections.
For details please see Reynolds, E.S., J. Cell Bio., 17, 208 (1963) and Sato, T., J. Electron Micro., 116, 133 (1976).
Venable, J.H. and Coggeshall, R. (1965). A simplified lead citrate stain for use in electron microscopy. J. Cell Biol., 25:407.
Famy, A. (1967). An extemporaneous lead citrate for electron microscopy. Proc. 25th Ann. EMSA Meet., p.148. Claitor's Pub. Division, Baton Rouge, LA.
Formula:
Pb(C₆H₅O₇)₂·3H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150456 CAS Number: 512-26-5 EINECS: 208-141-1 UN: 2291 ADR: 6.1,III |
Specification Test Results
Minimum assay | 99.% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...