Lead(II) acetate trihydrate ACS

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Acetic acid lead (II) salt trihydrate, Lead diacetate trihydrate

17600 17600-25
100504-068EA 45.64 USD
100504-068 100496-528
Lead(II) acetate trihydrate ACS
Lead(II) acetate trihydrate

We are now offering Lead Acetate in 25 ml bottles to avoid its classification as a hazardous chemical for shipping purposes.

Formula: (H₃CCOO)₂Pb·3H₂O
MW: 379.34 g/mol
Melting Pt: 75 °C (167 °F)
Density: 1.2 g/cm³ (10.014 lbs/gal.)
Storage Temperature: Refrigerator
MDL Number: MFCD00150023
CAS Number: 6080-56-4
UN: 1616
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=351511a44fbb598a2d7fa1fa4ab395427b6168f1&vid=67e858e5f0dc4d04dd796541b3c0ad5aa73f8dc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0 - 103.0%
Chloride 5 ppm
Copper 0.002%
Insoluble Matters 0.01%
Iron 0.001%
Nitrate and Nitrite (as NO₃) To pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR